APO455829655 Apollo Scientific Ltd

3-Amino-4-hydroxy-5-nitrobenzene-1-sulphonic acid hydrate

  • MDL number MFCD00277203
  • Cas Number : 175278-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25108

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25108
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart