APO455829668 Apollo Scientific Ltd

4-(1H-Pyrazol-3-yl)benzoic acid

  • MDL number MFCD01569442
  • Cas Number : 208511-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(4-Carboxyphenyl)-1H-pyrazole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-1h-pyrazol-3-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-pyrazol-3-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart