APO455829669 Apollo Scientific Ltd

2,2-Dimethylpentan-3-one

  • MDL number MFCD00039918
  • Cas Number : 564-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Dimethyl-3-oxopentane, Ethyl tert-butyl ketone
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25198

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25198
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart