APO455829674 Apollo Scientific Ltd

5-(Pyridin-2-yl)-1,3-oxazole

  • MDL number MFCD00085160
  • Cas Number : 70380-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 95-98/0.2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25219

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25219
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart