APO455829676 Apollo Scientific Ltd

1-Ethyl-1H-imidazole

  • MDL number MFCD00055389
  • Cas Number : 7098-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -27
Density 0.995
Flash Point 91.5
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 104/262mm
Product Link https://store.apolloscientific.co.uk/product/1-ethyl-1h-imidazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-ethyl-1h-imidazole
pictograms GHS07
hstatements H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart