APO455829680 Apollo Scientific Ltd

2-Chloro-1H-indole-3-carboxaldehyde

  • MDL number MFCD00111620
  • Cas Number : 5059-30-3
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 211-212
Iupac Name 2-chloro-1H-indole-3-carbaldehyde
Inchi Code 1S/C9H6ClNO/c10-9-7(5-12)6-3-1-2-4-8(6)11-9/h1-5,11H
Inchi Key XYSSNBNFOBVMAU-UHFFFAOYSA-N

Description

Synonym 2-Chloro-3-formyl-1H-indole
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 211-212
Product Link https://store.apolloscientific.co.uk/product/2-chloro-1h-indole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-1h-indole-3-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart