APO455829681 Apollo Scientific Ltd

3-(Chloromethyl)-5-thien-3-yl-1,2,4-oxadiazole

  • MDL number MFCD00111629
  • Cas Number : 184970-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65/70
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25269

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25269
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart