APO455829686 Apollo Scientific Ltd

5-(5-Bromothien-2-yl)isoxazole

  • MDL number MFCD03618406
  • Cas Number : 138716-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-67
Synonym 2-Bromo-5-(isoxazol-5-yl)thiophene, 5-(5-Bromothien-2-yl)-1,2-oxazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25306

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25306
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart