APO455829687 Apollo Scientific Ltd

3-(3-Methyl-1H-indol-1-yl)propanoic acid

  • MDL number MFCD00052088
  • Cas Number : 57662-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-85
Synonym 1-(Carboxyethyl)-3-methyl-1H-indole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25308

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25308
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart