APO455829692 Apollo Scientific Ltd

5-Bromo-2-chloro-4-methyl-3-nitropyridine

  • MDL number MFCD06659505
  • Cas Number : 884495-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Synonym 5-Bromo-2-chloro-3-nitro-4-picoline
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38098

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38098
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart