APO455829695 Apollo Scientific Ltd

N-(1,3-Benzodioxol-5-yl)-2-chloroacetamide

  • MDL number MFCD00245737
  • Cas Number : 227199-07-7
  • Molecular Weight : 213.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 155-156
Iupac Name N-(1,3-benzodioxol-5-yl)-2-chloroacetamide
Inchi Code 1S/C9H8ClNO3/c10-4-9(12)11-6-1-2-7-8(3-6)14-5-13-7/h1-3H,4-5H2,(H,11,12)
Inchi Key HBIYZYYNHWRQMC-UHFFFAOYSA-N

Description

Synonym N-(Chloroacetyl)-3,4-(methylenedioxy)aniline, 5-[(Chloroacetyl)amino]-1,3-benzodioxole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 155-156
Product Link https://store.apolloscientific.co.uk/product/n-13-benzodioxol-5-yl-2-chloroacetamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-13-benzodioxol-5-yl-2-chloroacetamide
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart