APO455829698 Apollo Scientific Ltd

5-(Methylsulphonyl)thiophene-2-carboxylic acid

  • MDL number MFCD00173745
  • Cas Number : 60166-86-1
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 201-204
Iupac Name 5-(methylsulfonyl)-2-thiophenecarboxylic acid
Inchi Code 1S/C6H6O4S2/c1-12(9,10)5-3-2-4(11-5)6(7)8/h2-3H,1H3,(H,7,8)
Inchi Key SLWRTVINHWIGTK-UHFFFAOYSA-N

Description

Synonym 5-(Methylsulphonyl)-2-thenoic acid, 2-Carboxy-5-(methylsulphonyl)thiophene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 201-204
Product Link https://store.apolloscientific.co.uk/product/5-methylsulphonylthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylsulphonylthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart