APO455829702 Apollo Scientific Ltd

Methyl 3,5-dibromobenzoate

  • MDL number MFCD00082648
  • Cas Number : 51329-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Synonym 1,3-Dibromo-5-(methoxycarbonyl)benzene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-35-dibromobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-35-dibromobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart