APO455829709 Apollo Scientific Ltd

4-(1H-Pyrrol-1-yl)aniline

  • MDL number MFCD01934575
  • Cas Number : 52768-17-9
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80.7-81.2
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-1h-pyrrol-1-ylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-pyrrol-1-ylaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart