APO455829716 Apollo Scientific Ltd

3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole

  • MDL number MFCD00728735
  • Cas Number : 259683-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [3-(Chloromethyl)-1,2,4-oxadiazol-5-yl]cyclobutane
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25527

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=25527
Availability details for
Quantity
  • Availability
  • Add to Cart