APO455829724 Apollo Scientific Ltd

Pyridine-3,4-dicarboximide

  • MDL number MFCD00013439
  • Cas Number : 4664-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-235
Synonym 1H-Pyrrolo[3,4-c]pyridine-1,3(2H)-dione, Cinchomeronimide, 2,3-Dihydro-1,3-dioxo-1H-pyrrolo[3,4-c]pyridine
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/pyridine-34-dicarboximide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/pyridine-34-dicarboximide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart