APO455829726 Apollo Scientific Ltd

3-(2,5-Dimethylfur-3-yl)-3-oxopropanenitrile

  • MDL number MFCD00275552
  • Cas Number : 175276-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Synonym 3-(Cyanoacetyl)-2,5-dimethylfuran, 3-(2,5-Dimethylfur-3-yl)-3-oxopropionitrile
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25642

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25642
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart