APO455829727 Apollo Scientific Ltd

5-(4-Chlorophenyl)-2-methylfuran-3-carbonyl chloride

  • MDL number MFCD00275557
  • Cas Number : 175276-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25644

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25644
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart