APO455829729 Apollo Scientific Ltd

Ethyl 2-methyl-5-(3-nitrophenyl)-3-furoate

  • MDL number MFCD00277691
  • Cas Number : 175276-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 2-methyl-5-(3-nitrophenyl)furan-3-carboxylate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25648

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25648
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart