APO455829731 Apollo Scientific Ltd

5-(1-Chloro-2-methylprop-2-yl)-3-(3-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD01765337
  • Cas Number : 680216-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[5-(1-Chloro-2-methylprop-2-yl)-1,2,4-oxadiazol-3-yl]toluene, 1-[5-(1-Chloro-2-methylprop-2-yl)-1,2,4-oxadiazol-3-yl]-3-methylbenzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25678

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25678
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart