APO455829732 Apollo Scientific Ltd

5-{3-[(tert-Butylsulphonyl)methyl]-1,2,4-oxadiazol-5-yl}-2-chloropyridine

  • MDL number MFCD02090296
  • Cas Number : 680216-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[(tert-Butylsulphonyl)methyl]-5-(6-chloropyridin-3-yl)-1,2,4-oxadiazole, tert-Butyl {[5-(6-chloropyridin-3-yl)-1,2,4-oxadiazol-3-yl]methyl} sulphone
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25686

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25686
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart