APO455829736 Apollo Scientific Ltd

(2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methylisoxazol-4-yl]carbamate

  • MDL number MFCD00207908
  • Cas Number : 261622-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazol-4-yl]carbamate, 2,6-Dichloro-4-[({[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazol-4-yl]carbamoyl}oxy)methyl]pyridine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25718

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25718
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart