APO455829738 Apollo Scientific Ltd

3,5-Dichlorobenzyl chloride

  • MDL number MFCD00060278
  • Cas Number : 3290-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-37
Synonym 1-(Chloromethyl)-2,3-dichlorobenzene, alpha,3,5-Trichlorotoluene
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Fused
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25740

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25740
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart