APO455829740 Apollo Scientific Ltd

5-Hydroxy-4-isopropyl-3-sulphanyl-4H-1,2,4-triazole

  • MDL number MFCD00068125
  • Cas Number : 53065-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Hydroxy-4-isopropyl-4H-1,2,4-triazole-3-thiol, 4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol, 4-(Prop-2-yl)-5-sulphanyl-4H-1,2,4-triazol-3-ol
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25753

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25753
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart