APO455829745 Apollo Scientific Ltd

2-Amino-5-methoxy-1,3-benzothiazole

  • MDL number MFCD00464314
  • Cas Number : 54346-87-1
  • Molecular Weight : 180.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Methoxy-1,3-benzothiazol-2-amine, 2-Amino-5-methoxybenzo[d]thiazole
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-methoxy-13-benzothiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-methoxy-13-benzothiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart