APO455829753 Apollo Scientific Ltd

4-Methoxy-2-nitrobenzaldehyde

  • MDL number MFCD00024254
  • Cas Number : 22996-21-0
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-2-nitrobenzaldehyde
Inchi Code 1S/C8H7NO4/c1-13-7-3-2-6(5-10)8(4-7)9(11)12/h2-5H,1H3
Inchi Key KLTDQLIGNSBZPO-UHFFFAOYSA-N

Description

Synonym 4-Formyl-3-nitroanisole, 2-Formyl-5-methoxynitrobenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methoxy-2-nitrobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxy-2-nitrobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart