APO455829759 Apollo Scientific Ltd

Ethyl 5-(tert-butyl)isoxazole-3-carboxylate

  • MDL number MFCD00085057
  • Cas Number : 91252-54-9
  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-tert-butylisoxazole-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-tert-butylisoxazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart