APO455829768 Apollo Scientific Ltd

6-Formyl-2-(methylthio)nicotinonitrile

  • MDL number MFCD00173824
  • Cas Number : 175277-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-139
Synonym 3-Cyano-2-(methylthio)pyridine-6-carboxaldehyde, 6-Formyl-2-(methylsulphanyl)nicotinonitrile
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25894

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25894
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart