APO455829775 Apollo Scientific Ltd

N-(Chloroacetyl)-3,5-dichloroaniline

  • MDL number MFCD00084930
  • Cas Number : 33560-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Synonym 2-Chloro-N-(3,5-dichlorophenyl)acetamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25932

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25932
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart