APO455829780 Apollo Scientific Ltd

2-Chlorophenoxyacetamidine hydrochloride

  • MDL number MFCD00173805
  • Cas Number : 58403-03-5
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 165-167
Iupac Name 2-(2-chlorophenoxy)ethanimidamide hydrochloride
Inchi Code 1S/C8H9ClN2O.ClH/c9-6-3-1-2-4-7(6)12-5-8(10)11;/h1-5H,10-11H2;1H
Inchi Key FVWIQEQOQCDMPN-UHFFFAOYSA-N

Description

Synonym 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 165-167
Product Link https://store.apolloscientific.co.uk/product/2-chlorophenoxyacetamidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chlorophenoxyacetamidine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart