APO455829781 Apollo Scientific Ltd

2-Bromo-1-[3-(4-chlorophenyl)isoxazol-5-yl]ethan-1-one

  • MDL number MFCD00662759
  • Cas Number : 258506-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-155
Synonym 2-Bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethan-1-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25949

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25949
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart