APO455829782 Apollo Scientific Ltd

Methyl 6-chloropyridazine-3-carboxylate

  • MDL number MFCD08694876
  • Cas Number : 65202-50-8
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Chloro-3-(methoxycarbonyl)pyridazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-6-chloropyridazine-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-chloropyridazine-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart