APO455829787 Apollo Scientific Ltd

2-(Phenylthio)ethanimidamide hydrochloride

  • MDL number MFCD00100031
  • Cas Number : 84544-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-177
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25975

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25975
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart