APO455829789 Apollo Scientific Ltd

6-Methylthionicotinamide

  • MDL number MFCD00114709
  • Cas Number : 175277-57-3
  • Molecular Weight : 152.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 193-196
Iupac Name 6-methyl-3-pyridinecarbothioamide
Inchi Code 1S/C7H8N2S/c1-5-2-3-6(4-9-5)7(8)10/h2-4H,1H3,(H2,8,10)
Inchi Key VBHUDUMOCHYQRM-UHFFFAOYSA-N

Description

Synonym 6-Methylpyridine-3-thiocarboxamide, 3-Carbamothioyl-6-methylpyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 193-196
Product Link https://store.apolloscientific.co.uk/product/6-methylthionicotinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-methylthionicotinamide
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart