APO455829791 Apollo Scientific Ltd

Ethyl 3-(fur-2-yl)-3-oxopropanoate

  • MDL number MFCD00122473
  • Cas Number : 615-09-8
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-(2-furyl)-3-oxopropanoate
Inchi Code 1S/C9H10O4/c1-2-12-9(11)6-7(10)8-4-3-5-13-8/h3-5H,2,6H2,1H3
Inchi Key PKHYBBXBBXOGMZ-UHFFFAOYSA-N

Description

Synonym 2-(3-Ethoxy-3-oxopropanoyl)furan, 1-Ethoxy-3-(fur-2-yl)propane-1,3-dione
Flash Point 115-120/4mm
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-fur-2-yl-3-oxopropanoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-fur-2-yl-3-oxopropanoate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart