APO455829794 Apollo Scientific Ltd

3-Chlorobenzhydrazide

  • MDL number MFCD00014757
  • Cas Number : 1673-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-158
Synonym 3-Chlorobenzoic acid hydrazide, 3-Chlorobenzohydrazide
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26002

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26002
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart