APO455829798 Apollo Scientific Ltd

2-Bromo-5-iodopyrazine

  • MDL number MFCD08061602
  • Cas Number : 622392-04-5
  • Molecular Weight : 284.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 95-97
Iupac Name 2-bromo-5-iodopyrazine
Inchi Code 1S/C4H2BrIN2/c5-3-1-8-4(6)2-7-3/h1-2H
Inchi Key OHTQHZVNZWWYFD-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-iodo-1,4-diazine
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 95-97
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-iodopyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-iodopyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart