APO455829804 Apollo Scientific Ltd

2-Chloroisonicotinamide

  • MDL number MFCD00221401
  • Cas Number : 100859-84-5
  • Molecular Weight : 156.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 201-203
Iupac Name 2-chloroisonicotinamide
Inchi Code 1S/C6H5ClN2O/c7-5-3-4(6(8)10)1-2-9-5/h1-3H,(H2,8,10)
Inchi Key DEMJOLRJLACBRX-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 201-203
Product Link https://store.apolloscientific.co.uk/product/2-chloroisonicotinamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloroisonicotinamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart