APO455829806 Apollo Scientific Ltd

2-Chloro-4-{3-[(4-methylphenoxy)methyl]-1,2,4-oxadiazol-5-yl}pyridine

  • MDL number MFCD00833093
  • Cas Number : 680216-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-({[5-(2-Chloropyridin-4-yl)-1,2,4-oxadiazol-3-yl]methyl}oxy)toluene, 5-(2-Chloropyridin-4-yl)-3-[(4-methylphenoxy)methyl]-1,2,4-oxadiazole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26091

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26091
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart