APO455829807 Apollo Scientific Ltd

alpha,2,6-Trichlorobenzaldoxime

  • MDL number MFCD00221426
  • Cas Number : 6579-27-7
  • Molecular Weight : 224.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-87
Synonym 2,6-Dichloro-N-hydroxybenzenecarboximidoyl chloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/alpha26-trichlorobenzaldoxime

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/alpha26-trichlorobenzaldoxime
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart