APO455829808 Apollo Scientific Ltd

2-Chloroisonicotinohydrazide

  • MDL number MFCD00221428
  • Cas Number : 58481-04-2
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloroisonicotinohydrazide
Inchi Code 1S/C6H6ClN3O/c7-5-3-4(1-2-9-5)6(11)10-8/h1-3H,8H2,(H,10,11)
Inchi Key MZIIYNBBSHJOLD-UHFFFAOYSA-N

Description

Synonym 2-Chloropyridine-4-carbohydrazide, 2-Chloroisonicotinic acid hydrazide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloroisonicotinohydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloroisonicotinohydrazide
pictograms GHS07
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart