APO455829810 Apollo Scientific Ltd

2-Chloro-6-methyl-3-nitropyridine

  • MDL number MFCD03085820
  • Cas Number : 56057-19-3
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-74
Synonym 6-Chloro-5-nitro-2-picoline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-methyl-3-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-methyl-3-nitropyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart