APO455829815 Apollo Scientific Ltd

4-Methylbenzamidine hydrochloride

  • MDL number MFCD00040183
  • Cas Number : 6326-27-8
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-214
Synonym 4-Methylbenzenecarboximidamide hydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylbenzamidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylbenzamidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H334
Availability details for
Quantity
  • Availability
  • Add to Cart