APO455829822 Apollo Scientific Ltd

1-Chloroisoquinoline-8-boronic acid

  • MDL number MFCD18257024
  • Cas Number : 2096337-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Borono-1-chloroisoquinoline, 8-Borono-1-chloro-2-azanaphthalene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51415

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51415
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart