APO455829823 Apollo Scientific Ltd

3-Chloroisoquinoline-6-boronic acid

  • MDL number MFCD22199342
  • Cas Number : 1800483-72-0
  • Molecular Weight : 207.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-6-isoquinolinylboronic acid
Inchi Code 1S/C9H7BClNO2/c11-9-4-7-3-8(10(13)14)2-1-6(7)5-12-9/h1-5,13-14H
Inchi Key CDJPJGXLCGBLRD-UHFFFAOYSA-N

Description

Synonym 6-Borono-3-chloroisoquinoline, 6-Borono-3-chloro-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloroisoquinoline-6-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloroisoquinoline-6-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart