APO455829826 Apollo Scientific Ltd

1-Chloroisoquinoline-5-carbonitrile

  • MDL number MFCD18250893
  • Cas Number : 1231761-23-1
  • Molecular Weight : 188.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-5-isoquinolinecarbonitrile
Inchi Code 1S/C10H5ClN2/c11-10-9-3-1-2-7(6-12)8(9)4-5-13-10/h1-5H
Inchi Key CXFDSWMDWPVWKF-UHFFFAOYSA-N

Description

Synonym 1-Chloro-5-cyanoisoquinoline, 1-Chloro-5-cyano-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-chloroisoquinoline-5-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-chloroisoquinoline-5-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart