APO455829828 Apollo Scientific Ltd

1-Chloroisoquinoline-6-carboxylic acid

  • MDL number MFCD15526626
  • Cas Number : 1256787-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2-azanaphthalene-6-carboxylic acid, 6-Carboxy-1-chloroisoquinoline
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54515

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54515
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart