APO455829830 Apollo Scientific Ltd

5-Bromo-3-hydroxyisoquinoline

  • MDL number MFCD11040413
  • Cas Number : 1175271-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromoisoquinolin-3-ol, 5-Bromo-3-hydroxy-2-azanaphthalene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54519

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54519
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart