APO455829834 Apollo Scientific Ltd

8-Bromo-1-chloroisoquinoline

  • MDL number MFCD13193666
  • Cas Number : 1233025-78-9
  • Molecular Weight : 242.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-1-chloroisoquinoline
Inchi Code 1S/C9H5BrClN/c10-7-3-1-2-6-4-5-12-9(11)8(6)7/h1-5H
Inchi Key RWBOOZDHIKMEHN-UHFFFAOYSA-N

Description

Synonym 8-Bromo-1-chloro-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-1-chloroisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-1-chloroisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart