APO455829835 Apollo Scientific Ltd

1-Hydroxyisoquinoline-5-carbonitrile

  • MDL number MFCD22375645
  • Cas Number : 90947-07-2
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Cyano-1-hydroxyisoquinoline, 5-Cyano-1-hydroxy-2-azanaphthalene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-hydroxyisoquinoline-5-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-hydroxyisoquinoline-5-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart